Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DECR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA580549
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA580549 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA580549 Supplier Invitrogen™ Supplier No. PA580549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human DECR1.

Auxiliary enzyme of beta-oxidation. It participates in the metabolism of unsaturated fatty enoyl-CoA esters having double bonds in both even- and odd-numbered positions. Catalyzes the NADP-dependent reduction of 2,4-dienoyl-CoA to yield trans-3-enoyl-CoA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DECR1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene DECR1
Gene Accession No. Q16698
Gene Alias 1200012F07Rik; 2,4-dienoyl CoA reductase 1, mitochondrial; 2,4-dienoyl-CoA reductase [NADPH]; 2,4-dienoyl-CoA reductase 1, mitochondrial; 2,4-dienoyl-CoA reductase, mitochondrial; 4-enoyl-CoA reductase; 4-enoyl-CoA reductase [NADPH]; Decr; DECR1; Nadph; SDR18C1; Short chain dehydrogenase/reductase family 18C member 1; short chain dehydrogenase/reductase family 18C, member 1
Gene Symbols DECR1
Host Species Rabbit
Immunogen E. coli-derived Human DECR1 fragment.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1666
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.