Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DECR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550080
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550080 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550080 Supplier Invitrogen™ Supplier No. PA550080
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total DECR2 protein.

DECR2 (2, 4-dienoyl-CoA reductase 2), also known as PDCR (peroxisomal 2, 4-dienoyl-CoA reductase) or SDR17C1, is a 292 amino acid member of the short-chain dehydrogenases/reductases (SDR) protein family and the 2, 4-dienoyl-CoA reductase protein subfamily. Localized to the peroxisome, DECR2 is an auxiliary enzyme of beta-oxidation that catalyzes the NADP-dependent reduction of 2, 4-dienoyl-CoA to yield trans-3-enoyl-CoA. DECR2 has also been shown to have catalytic activity towards 2, 4, 7, 10, 13, 16, 19-docosaheptaenoyl-CoA and short and medium chain 2, 4-dienoyl-CoAs, suggesting that DECR2 is not a rate limiting step in the degradation of docosahexaenoic acid in the peroxisome. DECR2 is expressed as three isoforms produced by alternative splicing events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DECR2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DECR2
Gene Accession No. Q9NUI1, Q9WV68
Gene Alias 2,4-dienoyl CoA reductase 2, peroxisomal; 2,4-dienoyl-CoA reductase 2; 2,4-dienoyl-CoA reductase 2, peroxisomal; 2-4-dienoyl-Coenzyme A reductase 2, peroxisomal; Dcrakl; DCR-AKL; Decr2; Pdcr; Peroxisomal 2,4-dienoyl-CoA reductase; putative peroxisomal 2,4-dienoyl-CoA reductase; putative peroxisomal 24-dienoyl-CoA reductase; putativeperoxisomal24-dienoyl-CoAreductase; pVI-AKL; SDR17C1; Short chain dehydrogenase/reductase family 17C member 1; short chain dehydrogenase/reductase family 17C, member 1
Gene Symbols DECR2
Host Species Rabbit
Immunogen Synthesized peptide derived from internal of human DECR2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26063, 26378
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.