Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dectin-1/CLEC7A Antibody (010), mFluor Violet 610 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290491MFV610
Description
Dectin-1/CLEC7A Monoclonal antibody specifically detects Dectin-1/CLEC7A in Mouse samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
Dectin-1/CLEC7A | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Immunocytochemistry | |
010 | |
Immunocytochemistry/ Immunofluorescence | |
Beta-glucan receptor, BGR, CLECSF12DC-associated C-type lectin 1, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 12, C-type lectin domain family 7 member A, C-type lectin domain family 7, member A, C-type lectin superfamily member 12, dectin-1, DECTIN1CANDF4, Dendritic cell-associated C-type lectin 1, dendritic cell-associated C-type lectin-1, hDectin-1, lectin-like receptor 1 | |
Rabbit immunized with purified, recombinant Mouse Dectin-1/CLEC7A (Accession#: NP_064392.2, Phe69-Leu244) | |
0.1 mL | |
Signal Transduction | |
64581 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction