Learn More
Invitrogen™ Dectin-1 Polyclonal Antibody

Description
Immunogen sequence: SWDGSKRQCW QLGSNLLKID SSNELGFIVK QVSSQPDNSF WIGLSRPQTE VPWLWEDGST FSSNLFQIRT TATQENPSPN CVWIHVSVIY DQLCSVPSYS I Positive Samples: H-460, SGC-7901, HL-60, Mouse spleen, Mouse thymus, Mouse liver, Mouse lung, Rat spleen; Cellular Location: Cell membrane, Cytoplasm, Single-pass type II membrane protein.
Specifications
Specifications
| Antigen | Dectin-1 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.61 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Clec7a |
| Gene Accession No. | Q6QLQ4, Q9BXN2 |
| Gene Alias | Beta-glucan receptor; beta-GR; BGR; CANDF4; CD369; CLEC7A; Clec7b; CLECSF12; C-type (calcium dependent, carbohydrate recognition domain) lectin, superfamily member 12; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 12; C-type lectin domain family 7 member A; C-type lectin domain family 7, member A; C-type lectin superfamily member 12; DC-associated C-type lectin 1; DECTIN1; dectin-1; Dendritic cell-associated C-type lectin 1; dendritic cell-associated C-type lectin-1; lectin-like receptor 1; RGD1565140; SCARE2; UNQ539/PRO1082 |
| Gene Symbols | Clec7a |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.