Learn More
Invitrogen™ Dectin 1 (soluble) Monoclonal Antibody (2A11), PE
Rat Monoclonal Antibody
$296.00 - $636.00
Specifications
Antigen | Dectin 1 (soluble) |
---|---|
Clone | 2A11 |
Concentration | Conc. Not Determined |
Content And Storage | 4°C, store in dark, DO NOT FREEZE! |
Applications | Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA516482
|
Invitrogen™
MA516482 |
100 Tests |
Each for $636.00
|
|
|||||
PIMA516483
|
Invitrogen™
MA516483 |
25 Tests |
Each for $296.00
|
|
|||||
Description
Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4ºC in the dark. Once reconstituted, this product has a shelf life of approx. 18 months. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Rat anti Mouse Dectin-1 antibody, clone 2A11 recognizes murine beta-glucan receptor, also known as Dectin-1. Rat anti Mouse Dectin-1 antibody, clone 2A11 recognizes murine beta-glucan receptor, also known as Dectin-1.
CLEC7A, also known as dectin-1, is a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily and is predominantly expressed on myeloid cells. It is a small glycoprotein type II membrane receptor with an extracellular C-type lectin-like domain fold and a cytoplasmic domain with an immunoreceptor tyrosine-based activation motif (ITAM). CLEC7A functions as a pattern-recognition receptor that recognizes a variety of beta-1,3-linked and beta-1,6-linked glucans from fungi and plants, and in this way plays a role in innate immune response. Upon fungal exposure, CLEC7A activates Syk tyrosine kinase, triggering a massive oxidative burst through the formation of reactive oxygen species.Specifications
Dectin 1 (soluble) | |
Conc. Not Determined | |
Flow Cytometry | |
PE | |
Rat | |
RUO | |
PBS with 1% BSA, 5% sucrose and 0.09% sodium azide | |
Q6QLQ4 | |
56644 | |
Dectin-1 transfected NIH3T3 cells and recombinant soluble Dectin-1. | |
Antibody |
2A11 | |
4°C, store in dark, DO NOT FREEZE! | |
Monoclonal | |
Lyophilized | |
IgG2b | |
Mouse | |
Clec7a | |
Beta-glucan receptor; beta-GR; BGR; CANDF4; CD369; CLEC7A; Clec7b; CLECSF12; C-type (calcium dependent, carbohydrate recognition domain) lectin, superfamily member 12; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 12; C-type lectin domain family 7 member A; C-type lectin domain family 7, member A; C-type lectin superfamily member 12; DC-associated C-type lectin 1; DECTIN1; dectin-1; Dendritic cell-associated C-type lectin 1; dendritic cell-associated C-type lectin-1; lectin-like receptor 1; RGD1565140; SCARE2; UNQ539/PRO1082 | |
Clec7a | |
Primary | |
Protein G |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.