Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dectin-2/CLEC6A Antibody (003), DyLight 755 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290507IR
Description
Dectin-2/CLEC6A Monoclonal antibody specifically detects Dectin-2/CLEC6A in Mouse samples. It is validated for ELISA, ELISASpecifications
| Dectin-2/CLEC6A | |
| Monoclonal | |
| DyLight 755 | |
| CLEC4N, CLECSF10, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 10, C-type lectin domain family 6 member A, C-type lectin domain family 6, member A, C-type lectin superfamily member 10, DC-associated C-type lectin 2, dectin 2, Dectin-2, DECTIN2, Dendritic cell-associated C-type lectin 2 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Dectin-2/CLEC6A (NP_064385.1; Ile44-Leu209). | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| ELISA | |
| 003 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 93978 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction