Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dectin-2/CLEC6A Antibody (1), Alexa Fluor™ 700, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290506AF700
Description
Dectin-2/CLEC6A Monoclonal specifically detects Dectin-2/CLEC6A in Mouse samples. It is validated for ELISA.Specifications
Dectin-2/CLEC6A | |
Monoclonal | |
Alexa Fluor 700 | |
CLEC4N, CLECSF10, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 10, C-type lectin domain family 6 member A, C-type lectin domain family 6, member A, C-type lectin superfamily member 10, DC-associated C-type lectin 2, dectin 2, Dectin-2, DECTIN2, Dendritic cell-associated C-type lectin 2 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Dectin-2/CLEC6A extracellular domain (NP_064385.1; Ile 44-Leu 209). | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
ELISA | |
1 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
93978 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction