Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dectin-2/CLEC6A Antibody (3D1) - Azide and BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$206.50 - $560.50

Specifications

Antigen Dectin-2/CLEC6A
Clone 3D1
Concentration 0.5mg/mL
Dilution Western Blot 2-5 ug/ml, Flow Cytometry 1ul/1 million cells, Immunohistochemistry-Paraffin 5 ug/ml, CyTOF-ready
Applications Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), CyTOF
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP236486SS
SDP
View Documents
Novus Biologicals
NBP236486SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP236486
SDP
View Documents
Novus Biologicals
NBP236486
0.1 mg
Each for $560.50
Only null left
Add to Cart
 
Description

Description

Dectin-2/CLEC6A Monoclonal specifically detects Dectin-2/CLEC6A in Human, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.
Specifications

Specifications

Dectin-2/CLEC6A
0.5mg/mL
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), CyTOF
Unconjugated
Mouse
Human
Q6EIG7
93978
A portion of amino acids 75-139 of human Dectin-2 was used as the immunogen for the antibody. 90-106
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
3D1
Western Blot 2-5 ug/ml, Flow Cytometry 1ul/1 million cells, Immunohistochemistry-Paraffin 5 ug/ml, CyTOF-ready
Monoclonal
Purified
RUO
PBS with No Preservative
CLEC4N, CLECSF10, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 10, C-type lectin domain family 6 member A, C-type lectin domain family 6, member A, C-type lectin superfamily member 10, DC-associated C-type lectin 2, dectin 2, Dectin-2, DECTIN2, Dendritic cell-associated C-type lectin 2
CLEC6A
IgG3 κ
Protein G purified
The amino acids sequence used as immunogen is 100% homologous in human, orangutan and chimp, 83% homologous in rhesus monkey, bovine and sheep, 65% homologous in mouse and 52% homologous in rat.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.