Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dectin-2/CLEC6A Antibody (3D1), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP227159PCP
Description
Dectin-2/CLEC6A Monoclonal specifically detects Dectin-2/CLEC6A in Human, Primate samples. It is validated for Flow Cytometry.Specifications
Dectin-2/CLEC6A | |
Monoclonal | |
PerCP | |
Q6EIG7 | |
CLEC6A | |
A portion of amino acids 75-139 of human Dectin-2 was used as the immunogen for the antibody. 90-106 | |
0.1 mL | |
Primary | |
The amino acids sequence used as immunogen is 100% homologous in Human, Orangutan and Chimp, 83% homologous in Rhesus Monkey, Bovine and Sheep, 65% homologous in Mouse and 52% homologous in Rat. | |
Store at 4C in the dark. | |
IgG3 κ |
Flow Cytometry, Immunohistochemistry (Paraffin) | |
3D1 | |
Flow Cytometry 1 μL/1 million cells | |
CLEC4N, CLECSF10, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 10, C-type lectin domain family 6 member A, C-type lectin domain family 6, member A, C-type lectin superfamily member 10, DC-associated C-type lectin 2, dectin 2, Dectin-2, DECTIN2, Dendritic cell-associated C-type lectin 2 | |
Mouse | |
Protein G purified | |
RUO | |
93978 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction