Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dectin-2/CLEC6A Antibody (RM0220-7B45) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$206.50 - $560.50
Specifications
Antigen | Dectin-2/CLEC6A |
---|---|
Clone | RM0220-7B45 |
Concentration | LYOPH |
Applications | Western Blot |
Classification | Monoclonal |
Description
Dectin-2/CLEC6A Monoclonal specifically detects Dectin-2/CLEC6A in Mouse samples. It is validated for Western Blot.Specifications
Dectin-2/CLEC6A | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
Q9JKF4 | |
93978 | |
IgG2 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
RM0220-7B45 | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
CLEC4N, CLECSF10, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 10, C-type lectin domain family 6 member A, C-type lectin domain family 6, member A, C-type lectin superfamily member 10, DC-associated C-type lectin 2, dectin 2, Dectin-2, DECTIN2, Dendritic cell-associated C-type lectin 2 | |
CLEC6A | |
Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title