Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Dectin 2 Monoclonal Antibody (3D1)
GREENER_CHOICE

Catalog No. PIMA516250
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516250 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516250 Supplier Invitrogen™ Supplier No. MA516250
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Sequence homology: Bovine/Sheep (83%), Mouse (65%), Rat (52%).

Dectin-2 is a C-type lectin expressed by dendritic cells, macrophages and inflammatory monocytes. It is a type II membrane receptor, with a short cytoplasmic tail, and a C-terminal C-type lectin domain. Dectin-2 is a receptor with specificity for high mannose structures and mediates cytokine production from DC and other antigen presenting cells. Though, Dectin-2 has no obvious signaling motifs attached to CP tail, reports indicate that Dectin-2 employs FcRg for internalization and to activate NF-kappaB pathway for cytokine production. Though it shares only 20-25% of homology with Dectin-1, it functions similarly as pattern recognition receptor (PRR) for fungal infections, to activate both innate (Syk-CARD9 innate signaling pathway) and adaptive immune responses (by activation of DC) to fungal infections. Thus Dectin-2 is a connector molecule of both innate and adaptive immunity functioning at DC level.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dectin 2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 3D1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene CLEC6A
Gene Accession No. Q6EIG7
Gene Alias Clec4n; CLEC6A; Clec6a-ps1; CLECSF10; C-type (calcium dependent, carbohydrate recognition domain) lectin, superfamily member 10; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 10; C-type lectin domain family 4, member n; C-type lectin domain family 6 member A; C-type lectin domain family 6, member A; C-type lectin domain family 6, member A, pseudogene 1; C-type lectin superfamily member 10; DC-associated C-type lectin 2; dectin 2; DECTIN2; dectin-2; Dectin-2p; Dendritic cell-associated C-type lectin 2; natural killer complex lectin; Nkcl
Gene Symbols CLEC6A
Host Species Mouse
Immunogen A blend of two peptides made to amino acids 44-63 and 90-106 of human Dectin-2.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93978
Target Species Human, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG3 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.