Learn More
Invitrogen™ Dectin 2 Recombinant Rabbit Monoclonal Antibody (003)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA529160
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Mouse CLEC4N/CLEC6A/Dectin-2.
Dectin-2 is a C-type lectin expressed by dendritic cells, macrophages and inflammatory monocytes. It is a type II membrane receptor, with a short cytoplasmic tail, and a C-terminal C-type lectin domain. Dectin-2 is a receptor with specificity for high mannose structures and mediates cytokine production from DC and other antigen presenting cells. Though, Dectin-2 has no obvious signaling motifs attached to CP tail, reports indicate that Dectin-2 employs FcRg for internalization and to activate NF-kappaB pathway for cytokine production. Though it shares only 20-25% of homology with Dectin-1, it functions similarly as pattern recognition receptor (PRR) for fungal infections, to activate both innate (Syk-CARD9 innate signaling pathway) and adaptive immune responses (by activation of DC) to fungal infections. Thus Dectin-2 is a connector molecule of both innate and adaptive immunity functioning at DC level.
Specifications
Dectin 2 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q9JKF4 | |
CLEC6A | |
Recombinant Mouse CLEC4N/CLEC6A/Dectin-2 Protein (Ile44-Leu209). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
ELISA | |
003 | |
Unconjugated | |
CLEC6A | |
Clec4n; CLEC6A; Clec6a-ps1; CLECSF10; C-type (calcium dependent, carbohydrate recognition domain) lectin, superfamily member 10; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 10; C-type lectin domain family 4, member n; C-type lectin domain family 6 member A; C-type lectin domain family 6, member A; C-type lectin domain family 6, member A, pseudogene 1; C-type lectin superfamily member 10; DC-associated C-type lectin 2; dectin 2; DECTIN2; dectin-2; Dectin-2p; Dendritic cell-associated C-type lectin 2; natural killer complex lectin; Nkcl | |
Rabbit | |
Protein A | |
RUO | |
56620 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.