Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decyl sodium sulfate, HPLC grade

Catalog No. p-3778123
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

Catalog No. AC406220050

Supplier: Thermo Scientific Chemicals 406220050

Only null left
Add to Cart

CAS: 142-87-0 | C10H19Na·H2SO4 | 260.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ion pair chromatography
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-87-0
MDL Number MFCD00041881
InChI Key XZTJQQLJJCXOLP-UHFFFAOYSA-M
Synonym sodium decyl sulfate, sodium n-decyl sulfate, decyl sodium sulfate, sulfuric acid, monodecyl ester, sodium salt, sodium decyl sulphate, unii-al92m833sy, n-decyl sodium sulfate, sodium n-decylsulphate, sulfuric acid, monodecyl ester, sodium salt 1:1, sulfuric acid, decyl ester, sodium salt
PubChem CID 23665771
IUPAC Name sodium;decyl sulfate
SMILES CCCCCCCCCCOS(=O)(=O)[O-].[Na+]

Specifications

Absorbance (0.005M in water) (1 cm cell), (0.005 M in water) (1 cm cell) at 0nm, 0.01 max. at 254nm, 0.01 max. at 280nm, 0.05 max. at 220nm, 0.1 max. at 200nm
Color White to Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C10H21NaO4S
Linear Formula CH3(CH2)9OSO3Na
Quantity 5 g
Molecular Weight (g/mol) 260.324
Formula Weight 260.32
Percent Purity ≥98.5% (titrimetric) (on dry substance)
Grade HPLC
Physical Form Powder
Chemical Name or Material Decyl sodium sulfate
Show More Show Less
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.