Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DEDD Polyclonal Antibody
DEDD Polyclonal Antibody
Supplier: Thermo Scientific PA583152
Description
DEDD Polyclonal Antibody for Western Blot, ICC/IF

Specifications
DEDD | |
Polyclonal | |
Unconjugated | |
DEDD | |
CASP8IP1; death effector domain containing; death effector domain-containing; death effector domain-containing protein; Death effector domain-containing testicular molecule; DEDD; DEDD1; Dedpro1; DEFT; FLDED1; FLDED-1; KE05; tumor necrosis factor (ligand) superfamily, member 19 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9191 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75618 | |
DEDD | |
Recombinant protein corresponding to Human DEDD. Recombinant protein control fragment (Product #RP-104613). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction