Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DEDD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584930
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584930 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584930 Supplier Invitrogen™ Supplier No. PA584930
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LALERQGRCD ESNFRQVLQL LRIITRHDLL PYVTLKRRRA VCPDLVDKYL EETSIRYVTP RALSD Highest antigen sequence indentity to the following orthologs: Mouse - 98%, Rat - 98%.

DEDD encodes a protein that contains a death effector domain (DED). DED is a protein-protein interaction domain shared by adaptors, regulators and executors of the programmed cell death pathway. Overexpression of this gene was shown to induce weak apoptosis. Upon stimulation, this protein was found to translocate from cytoplasm to nucleus and colocalize with UBTF, a basal factor required for RNA polymerase I transcription, in the nucleolus. At least three transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DEDD
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DEDD
Gene Accession No. O75618
Gene Alias CASP8IP1; death effector domain containing; death effector domain-containing; death effector domain-containing protein; Death effector domain-containing testicular molecule; DEDD; DEDD1; Dedpro1; DEFT; FLDED1; FLDED-1; KE05; tumor necrosis factor (ligand) superfamily, member 19
Gene Symbols DEDD
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DEDD. Recombinant protein control fragment (Product #RP-108616).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9191
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.