Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DEDD Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | DEDD |
---|---|
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
DEDD Polyclonal specifically detects DEDD in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
DEDD | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
CASP8IP1, death effector domain containing, death effector domain-containing, death effector domain-containing protein, Death effector domain-containing testicular molecule, DEDD1, DEDPRO1, DEFTKE05, FLDED1, FLDED-1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-318 of human DEDD (NP_001034801.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Apoptosis | |
PBS with 50% glycerol, pH7.3. | |
9191 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title