Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DEDD2 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056137T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056137T 0.025 mL
NB10056137 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056137T Supplier Novus Biologicals Supplier No. NB10056137SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DEDD2 Polyclonal specifically detects DEDD2 in Human, Rat, Bovine, Canine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen DEDD2
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000
Formulation Whole antisera with 0.02% Sodium Azide
Gene Alias death effector domain containing 2, death effector domain-containing DNA binding protein 2, DED-containing protein FLAME-3, DNA-binding death effector domain-containing protein 2, FADD-like apoptotic molecule 3, FLAME3, FLAME-3
Gene Symbols DEDD2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 210-228 (GPALEQGVASRRPQALARQ) of human DEDD2/Flame-3 was used as immunogen; GenBank no. gi 19923050 ref NP_579874.1 .
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 162989
Test Specificity DEDD2
Target Species Human, Rat, Bovine, Canine, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.