Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DEDD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572905
Description
DEDD2 Polyclonal Antibody for Western Blot, IHC (P), IP
Apoptotic signals are often triggered by cell surface death receptors through protein-protein interactions mediated by conserved domains such as the death effector domain. A novel death effector domain (DED)-containing protein, DEDD2, has been recently identified. Over-expression of DEDD2 in transfected cells induces moderate apoptosis and results in substantial sensitization to apoptosis induced by Fas, TRAIL, and FADD. More recently, it has been shown that DEDD2 can bind caspase-8 and -10 in addition to FLIP but not FADD. Like the related protein DEDD, DEDD2 translocates from the cytosol to the nucleus upon induction of apoptosis, and it has been suggested that DEDD2 may target caspase-8 to the nucleus and that DEDD2 thus plays a critical role in death receptor-induced apoptosis. At least two alternatively spliced transcript variants encoding distinct isoforms have been found for DEDD2.
Specifications
DEDD2 | |
Polyclonal | |
Unconjugated | |
Dedd2 | |
2410050E11Rik; death effector domain containing 2; death effector domain-containing DNA binding protein 2; death effector domain-containing DNA-binding protein 2; death effector domain-containingDNA binding protein 2; DED-containing protein FLAME-3; DEDD2; DNA-binding death effector domain-containing protein 2; FADD-like anti-apoptotic molecule 3; FLAME3; FLAME-3; PSEC0004 | |
Rabbit | |
50 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Conc. Not Determined | |
Whole serum with 0.02% sodium azide | |
Q8WXF8 | |
Dedd2 | |
A synthetic peptide corresponding to amino acids 175-193 of human DEDD2/Flame-3. | |
RUO | |
162989 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction