Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DEDD2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | DEDD2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
DEDD2 Polyclonal specifically detects DEDD2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
DEDD2 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
162989 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
death effector domain containing 2, death effector domain-containing DNA binding protein 2, DED-containing protein FLAME-3, DNA-binding death effector domain-containing protein 2, FADD-like apoptotic molecule 3, FLAME3, FLAME-3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human DEDD2 (NP_579874.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title