Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

dedicator of cytokinesis 8 Antibody (OTI12D7), HRP, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP272227H

Catalog No. NBP272227H


Only null left
Add to Cart

Description

Description

dedicator of cytokinesis 8 Monoclonal specifically detects dedicator of cytokinesis 8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

dedicator of cytokinesis 8
Monoclonal
HRP
PBS with No Preservative
DOCK8
Recombinant protein fragment corresponding to amino acids 84-405 of human dedicator of cytokinesis 8 (NP_982272) produced in E.coli.
0.1 mL
81704.0
Store at 4°C in the dark.
Western Blot, Immunohistochemistry
OTI12D7
Western Blot, Immunohistochemistry
dedicator of cytokinesis 8, FLJ00026, FLJ00346
Mouse
Affinity Purified
Primary
Human, Mouse, Rat
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.