Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
dedicator of cytokinesis 8 Antibody (OTI4F3), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272226AF700
Description
dedicator of cytokinesis 8 Monoclonal specifically detects dedicator of cytokinesis 8 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
dedicator of cytokinesis 8 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
DOCK8 | |
Full length recombinant protein of human dedicator of cytokinesis 8 (NP_982272) produced in HEK293T cells. | |
0.1 mL | |
81704.0 | |
Store at 4°C in the dark. |
Immunohistochemistry | |
OTI4F3 | |
Immunohistochemistry | |
dedicator of cytokinesis 8, FLJ00026, FLJ00346 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction