Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
dedicator of cytokinesis 8 Antibody (OTI4F3), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272226PCP
Description
dedicator of cytokinesis 8 Monoclonal antibody specifically detects dedicator of cytokinesis 8 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
dedicator of cytokinesis 8 | |
Monoclonal | |
PerCP | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
OTI4F3 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
dedicator of cytokinesis 8, FLJ00026, FLJ00346 | |
Full length human recombinant protein of human DOCK8(NP_982272) produced in HEK293T cell. | |
0.1 mL | |
Cardiovascular Biology, Signal Transduction | |
81704 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction