Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
dedicator of cytokinesis 8 Antibody (OTI4H10), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272228UV
Description
dedicator of cytokinesis 8 Monoclonal specifically detects dedicator of cytokinesis 8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
dedicator of cytokinesis 8 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
DOCK8 | |
Recombinant protein fragment corresponding to amino acids 832-1160 of human dedicator of cytokinesis 8 (NP_982272) produced in E.coli. | |
0.1 mL | |
81704.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI4H10 | |
Western Blot, Immunohistochemistry | |
dedicator of cytokinesis 8, FLJ00026, FLJ00346 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction