Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DEF6 Antibody (OTI3F9), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271868H
Description
DEF6 Monoclonal specifically detects DEF6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
DEF6 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
DEF6 | |
Full length recombinant protein of human DEF6 (NP_071330) produced in HEK293T cells. | |
0.1 mL | |
50619.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI3F9 | |
Western Blot, Immunohistochemistry | |
DEF-6, differentially expressed in FDCP 6 homolog, differentially expressed in FDCP 6 homolog (mouse), IBPdifferentially expressed in FDCP (mouse homolog) 6, IRF4-binding protein, SLAT, SWAP70L, SWAP-70-like adaptor protein of T cells | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction