Learn More
Invitrogen™ DEF6 Monoclonal Antibody (2D2)
Mouse Monoclonal Antibody
Supplier:  Invitrogen™ MA524389
Catalog No. PIMA524389
Only null left
Add to Cart
Description
Description
Peptide Sequence: MRTLTILTAV LLVALQAKAE PLQAEDDPLQ AKAYEADAQE QRGANDQDFA VSFAEDASSS LRALGSTRAF TCHCRRSCYS TEYSYGTCTV MGINHRFCCL.
DEF6, or IBP, is a guanine nucleotide exchange factor for RAC and CDC42 that is highly expressed in B and T cells.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DEF6
Monoclonal
Unconjugated
DEF6
2410003F05Rik; 6430538D02Rik; AV094905; DEF6; DEF-6; DEF6 guanine nucleotide exchange factor; DEF6, guanine nucleotide exchange factor; differentially expressed in FDCP 6; Differentially expressed in FDCP 6 homolog; differentially expressed in FDCP 6 homolog (mouse); Ibp; IRF-4-binding protein; IRF4-binding protein; LOW QUALITY PROTEIN: differentially expressed in FDCP 6 homolog; OTTHUMP00000016251; RP3-329A5.5; SLAT; Slat2; Slat6; SWAP70L; SWAP-70-like adapter of T-cells; SWAP-70-like adaptor protein of T cells
Mouse
Protein A
RUO
50619
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
2D2
PBS with no preservative; pH 7.4
Q9H4E7
DEF6
DEFA6 (NP_001917.1, 1 a.a. ~ 100 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG2a κ
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.