Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DEF6 Monoclonal Antibody (OTI2C11), TrueMAB™, OriGene
SDP

Catalog No. 502539777
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-253-9777 30 μL
50-167-5821 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-253-9777 Supplier OriGene Supplier No. TA505324S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DEF6 Monoclonal antibody specifically detects DEF6 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen DEF6
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2C11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene DEF6
Gene Accession No. Q9H4E7
Gene Alias 2410003F05Rik; 6430538D02Rik; AV094905; DEF6; DEF-6; DEF6 guanine nucleotide exchange factor; DEF6, guanine nucleotide exchange factor; differentially expressed in FDCP 6; Differentially expressed in FDCP 6 homolog; differentially expressed in FDCP 6 homolog (mouse); Ibp; IRF-4-binding protein; IRF4-binding protein; LOW QUALITY PROTEIN: differentially expressed in FDCP 6 homolog; OTTHUMP00000016251; RP3-329A5.5; SLAT; Slat2; Slat6; SWAP70L; SWAP-70-like adapter of T-cells; SWAP-70-like adaptor protein of T cells
Host Species Mouse
Immunogen Full length human recombinant protein of human DEF6 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50619
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.