Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DEF6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545672
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545672 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545672 Supplier Invitrogen™ Supplier No. PA545672
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: QERYIERAQQ EKEELQQEMA QQSRSLQQAQ QQLEEVRQNR QRADEDVEAA Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 83%; Horse: 93%; Human: 100%; Mouse: 93%; Pig: 90%; Rabbit: 93%; Rat: 93%.

DEF6, or IBP, is a guanine nucleotide exchange factor for RAC and CDC42 that is highly expressed in B and T cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DEF6
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DEF6
Gene Accession No. Q9H4E7
Gene Alias 2410003F05Rik; 6430538D02Rik; AV094905; DEF6; DEF-6; DEF6 guanine nucleotide exchange factor; DEF6, guanine nucleotide exchange factor; differentially expressed in FDCP 6; Differentially expressed in FDCP 6 homolog; differentially expressed in FDCP 6 homolog (mouse); Ibp; IRF-4-binding protein; IRF4-binding protein; LOW QUALITY PROTEIN: differentially expressed in FDCP 6 homolog; OTTHUMP00000016251; RP3-329A5.5; SLAT; Slat2; Slat6; SWAP70L; SWAP-70-like adapter of T-cells; SWAP-70-like adaptor protein of T cells
Gene Symbols DEF6
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human DEF6 (aa 480-529).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50619
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.