Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DEF8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559706
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559706 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559706 Supplier Invitrogen™ Supplier No. PA559706
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HNWDFEPRKV SRCSMRYLAL MVSRPVLRLR EINPLLFSYV EELVEIRKLR QDILLMKPYF ITCREAMEAR LLLQLQDRQH FVENDEMYSV QD Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

Positively regulates lysosome peripheral distribution and ruffled border formation in osteoclasts. Involved in bone resorption. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DEF8
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DEF8
Gene Accession No. Q6ZN54
Gene Alias DEF8; DEF-8; Differentially expressed in FDCP 8 homolog; differentially expressed in FDCP 8 homolog (mouse)
Gene Symbols DEF8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DEF8. Recombinant protein control fragment (Product #RP-98690).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54849
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.