Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ DEFA1, Rabbit, Unlabeled, Polyclonal,

Catalog No. ABF1159MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABF1159MI 50 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABF1159MI Supplier MilliporeSigma™ Supplier No. ABF1159
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The human DEFA1 gene (also known as DEF1, DEFA2, MRS, DEFA1B; Gene ID 1667) encodes a 94-amino acid precursor protein (UniProt P59665) with an N-terminal signal peptide (a.a. 1-19) and propeptide (a.a. 20-38) sequence. Posttranslational processing by proteolytic cleavages generates three neutrophil granule peptides, HP 1-56 (a.a. 39-94), neutrophil defensin 1 (HNP-1, HP-1, HP1; a.a. 65-94), and neutrophil defensin 2 (HNP-2, HP-2, HP2; a.a. 66-94). These peptides exhibit several biological activities including the non-oxidative destruction of ingested microorganisms, the inhibition of adrenocorticotropin-stimulated glucocorticoids synthesis, monocyte chemotaxis, the non-cytolytic inhibition of thymidine incorporation in HL-60 promyelocyte-like cells and the stimulation of nifedipine-sensitive calcium channels. The richest source of HP1-56 relative to HP-1 is found in fetal lung tissue. In addition, HP1-56 is found in the HL-60 promyelocytic cell line and in some leukemias, but not in normal mature circulating neutrophils.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DEFA1
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene DEFA1, DEF1, DEFA2, MRS, DEFA1B
Gene Accession No. P59665
Host Species Rabbit
Immunogen KLH-conjuated linear peptide corresponding to an internal sequence of human HP 1-56.
Purification Method Affinity Purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_004075
Test Specificity This rabbit polyclonal antibody targets a region present in prepro-, pro-, and mature HP 1-56 (a.a. 1-94, 20-94, and 39-94, respectively). Target region is absent from neutrophil defensin 1 (HNP-1, HP-1, HP1; a.a. 65-94) and neutrophil defensin 2 (HNP-2, HP-2, HP2; a.a. 66-94).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.