Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DEFA4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562362
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562362 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562362 Supplier Invitrogen™ Supplier No. PA562362
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSALQVSGST RGMVCSCRLV FCRRTELRVG NCLIGGVSFT YCCTRVD Highest antigen sequence identity to the following orthologs: Mouse - 32%, Rat - 36%.

Defensins are a family of antimicrobial and cytotoxic peptides thought to be involved in host defense. They are abundant in the granules of neutrophils and also found in the epithelia of mucosal surfaces such as those of the intestine, respiratory tract, urinary tract, and vagina. Members of the defensin family are highly similar in protein sequence and distinguished by a conserved cysteine motif. Several alpha defensin genes are clustered on chromosome 8. This gene differs from other genes of this family by an extra 83-base segment that is apparently the result of a recent duplication within the coding region. The protein encoded by this gene, defensin, alpha 4, is found in the neutrophils; it exhibits corticostatic activity and inhibits corticotropin stimulated corticosterone production.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DEFA4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DEFA4
Gene Accession No. P12838
Gene Alias Alpha-defensin 4; corticostatin; Crp4; DEF4; Defa28; Defa4; Defcr4; defensin alpha 4; Defensin, alpha 4; defensin, alpha 4, corticostatin; defensin, alpha 4, preproprotein; defensin, alpha, 4; Defensin-related cryptdin-4; HNP-4; HP4; HP-4; Neutrophil defensin 4
Gene Symbols DEFA4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Defensin. Recombinant protein control fragment (Product #RP-101423).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1669
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.