Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Defactinib
GREENER_CHOICE

Catalog No. 730525
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
73-052-5 25 mg
73-055 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 73-052-5 Supplier Tocris Bioscience™ Supplier No. 7305/25
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective FAK and Pyk2 inhibitor

Defactinib is a potent and selective focal adhesion kinase (FAK) and proline rich tyrosine kinase 2 (Pyk2) inhibitor (IC50 = 0.6 nM at both). The compound shows >100-fold selectivity for FAK and Pyk2 over other kinases. Defactinib inhibits FAK phosphorylation in vivo (EC50 = 26 nM), and inhibits tumor growth. Defactinib also decreases the viability of thyroid cancer cells, showing greater selectivity for TT and K1 cells compared to FAK Inhibitor 14 (Cat. No. 3414) (IC50 values are 1.98 μM and 10.34 μM, respectively). Defactinib suppresses colony growth of Pnf cells when combined with Selumetinib (Cat. No. 6815).

Specifications

Chemical Name or Material N-Methyl-4-[[4-[[[3-[methyl(methylsulfonyl)amino]-2-pyrazinyl]methyl]amino]-5-(trifluoromethyl)-2-pyrimidinyl]amino]benzamide
CAS 1073154-85-4
Quantity 25 mg
Target Focal Adhesion Kinase Inhibitors
Molecular Formula C20H21F3N8O3S
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.