Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DEFB1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | DEFB1 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-9684
![]() ![]() |
Proteintech Group Inc
147381AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Beta-defensins (also designated BD, and hBD in human) are small cationic peptides with broad-spectrum antimicrobial activity. Produced in mucosal epithelia and neutrophils of several species, beta-defensins are developmentally regulated. Research indicates that human beta-defensin 1 may contribute to the antimicrobial activity of airway surface fluid and may play a role in the mucosal defenses of the lung. Human beta-defensin 1 shares homology with other beta-defensins from human blood filtrate and is also present in omolar concentrations in human plasma. In addition to the antimicrobial activity of human airway epithelia, beta-defensin 1 may play a role in the mucosal defenses of the lung.Specifications
DEFB1 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
P60022 | |
1672 | |
DEFB1 Fusion Protein Ag6406 | |
Primary | |
-20°C | |
DEFB1 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
BD 1, BD1, Beta defensin 1, DEFB 1, DEFB1, DEFB101, defensin, beta 1, hBD 1, HBD1 | |
DEFB1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title