Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DEFB104A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583768
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583768 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583768 Supplier Invitrogen™ Supplier No. PA583768
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EFELDRICGY GTARCRKKCR SQEYRIGRCP NTYACCLRKW DESLLNRT Highest antigen sequence indentity to the following orthologs: Rat - 33%, Mouse - 33%.

Defensins form a family of antimicrobial and cytotoxic peptides made by neutrophils. Defensins are short, processed peptide molecules that are classified by structure into three groups: alpha-defensins, beta-defensins and theta-defensins. All beta-defensin genes are densely clustered in four to five syntenic chromosomal regions. Chromosome 8p23 contains at least two copies of the duplicated beta-defensin cluster. This duplication results in two identical copies of defensin, beta 104, DEFB104A and DEFB104B, in head-to-head orientation. This gene, DEFB104A, represents the more centromeric copy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DEFB104A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DEFB104A
Gene Accession No. Q8WTQ1
Gene Alias BD-4; Beta-defensin 104; Beta-defensin 4; DEFB104; DEFB104A; DEFB104B; DEFB4; DEFB-4; defensin beta 104A; Defensin, beta 104; defensin, beta 104A; defensin, beta 4; hBD-4
Gene Symbols DEFB104A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DEFB104A. Recombinant protein control fragment (Product #RP-108181).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140596
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.