Learn More
DEFB104A Polyclonal Antibody, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
$319.00 - $2175.00
Specifications
Antigen | DEFB104A |
---|---|
Concentration | 0.1-1.0 mg/mL |
Applications | ELISA, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
500P268100
![]() |
Invitrogen™
500P268100UG |
100 μg |
Each for $319.00
|
|
|||||
500P2681MG
![]() |
Invitrogen™
500P2681MG |
1 mg |
Each for $2,175.00
|
|
|||||
Description
AA Sequence of recombinant protein: EFELDRICGY GTARCRKKCR SQEYRIGRCP NTYACCLRKW DESLLNRTKP. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human BD-4. Anti-Human BD-4-specific antibody was purified by affinity chromatography employing an immobilized Human BD-4 matrix. Sandwich ELISA: To detect Human BD-4 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human BD-4 (500-P268Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human BD-4. Western Blot: To detect Human BD-4 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human BD-4 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
Defensins form a family of antimicrobial and cytotoxic peptides made by neutrophils. Defensins are short, processed peptide molecules that are classified by structure into three groups: alpha-defensins, beta-defensins and theta-defensins. All beta-defensin genes are densely clustered in four to five syntenic chromosomal regions. Chromosome 8p23 contains at least two copies of the duplicated beta-defensin cluster. This duplication results in two identical copies of defensin, beta 104, DEFB104A and DEFB104B, in head-to-head orientation. This gene, DEFB104A, represents the more centromeric copy.Specifications
DEFB104A | |
ELISA, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q8WTQ1 | |
140596 | |
E.coli-derived Recombinant Human BD-4. | |
Antigen affinity chromatography | |
DEFB104A |
0.1-1.0 mg/mL | |
Polyclonal | |
Lyophilized | |
RUO | |
PBS with no preservative | |
BD-4; Beta-defensin 104; Beta-defensin 4; DEFB104; DEFB104A; DEFB104B; DEFB4; DEFB-4; defensin beta 104A; Defensin, beta 104; defensin, beta 104A; defensin, beta 4; hBD-4 | |
DEFB104A | |
Primary | |
-20°C |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.