Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DEFB107A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | DEFB107A |
---|---|
Dilution | Immunohistochemistry-Paraffin 1:50 - 1:200 |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
DEFB107A Polyclonal specifically detects DEFB107A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
DEFB107A | |
Polyclonal | |
Purified | |
RUO | |
BD-7, Beta-Defensin 107, Beta-Defensin 7, DEFB107, DEFB107A DEFB107B, DEFB7, DEFB-7, Defensin Beta 107A, Defensin, Beta 107A, Defensin, Beta 7 | |
DEFB107A | |
IgG | |
Protein A purified |
Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Unconjugated | |
Rabbit | |
Human | |
245910 | |
This antibody was developed against a recombinant protein corresponding to the amino acid sequence:QARTAIHRALISKRMEGHCEAECLTFEVKIGGCRAELAP | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title