Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DEFB112 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548687
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548687 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548687 Supplier Invitrogen™ Supplier No. PA548687
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DEFB112 Polyclonal Antibody for Western Blot, Flow

Has antibacterial activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DEFB112
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene DEFB112
Gene Accession No. Q30KQ8
Gene Alias beta-defensin 112; beta-defensin 12; DEFB112; DEFB-12; defensin beta 112; defensin, beta 112; defensin, beta 12
Gene Symbols DEFB112
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 15-43 amino acids from the N-terminal region of human DEFB112.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 245915
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.