Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DEFB118 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560061
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560061 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560061 Supplier Invitrogen™ Supplier No. PA560061
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KCWNRSGHCR KQCKDGEAVK DTCKNLRACC IPSNEDHRRV PATSPTPLSD STPGIIDDIL TVRFTTDYFE VSSKKDMVEE SEAGRGTETS LPNVHHSS Highest antigen sequence identity to the following orthologs: Mouse - 23%, Rat - 25%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen DEFB118
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DEFB118
Gene Accession No. Q96PH6
Gene Alias beta-defensin 118; Beta-defensin 18; C20orf63; DEFB118; DEFB18; DEFB-18; defensin beta 118; defensin, beta 118; defensin, beta 18; epididymal secretory protein 13.6; epididymus specific clone 42; ESC42; ESP13.6
Gene Symbols DEFB118
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DEFB118. Recombinant protein control fragment (Product #RP-99426).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117285
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.