Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DEFB119 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560222
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560222 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560222 Supplier Invitrogen™ Supplier No. PA560222
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KRHILRCMGN SGICRASCKK NEQPYLYCRN CQSCCLQSYM RISISGKEEN TDWSYEKQWP RLP Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 65%.

This gene encodes a member of the beta subfamily of defensins. Beta-defensins are antimicrobial peptides that protect tissues and organs from infection by a variety of microorganisms. This gene is found in a cluster with other beta-defensin genes on the long arm of chromosome 20. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DEFB119
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DEFB119
Gene Accession No. Q8N690
Gene Alias Beta-defensin 119; beta-defensin 120; Beta-defensin 19; Beta-defensin 20; CBD119; CBD120; DEFB119; DEFB120; DEFB19; DEFB-19; DEFB20; DEFB-20; defensin beta 119; defensin, beta 119; Defensin, beta 120; defensin, beta 19; defensin, beta 20; ESC42-RELA; ESC42-RELB; UNQ2449/PRO5729
Gene Symbols DEFB119
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DEFB119. Recombinant protein control fragment (Product #RP-99456).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 245932
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.