Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DEFB129 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582507
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582507 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582507 Supplier Invitrogen™ Supplier No. PA582507
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CCVGPKVVKL IKNYLQYGTP NVLNEDVQEM LKPAKNSSAV IQRKHILSVL PQIKSTSFFA NTNFVIIPNA TPMNSATIST MTPGQITYTA TSTKSNTKES RDSATASPPP APPPPNILPT PSLEL Highest antigen sequence indentity to the following orthologs: Rat - 36%, Mouse - 39%.

DEFB129 gene ontology annotations related to this gene include innate immune response.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DEFB129
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DEFB129
Gene Accession No. Q9H1M3
Gene Alias bA530N10.3; BD129; beta defensin 29; beta-defensin 129; Beta-defensin 29; C20orf87; CBD129; DEFB129; DEFB29; DEFB-29; defensin beta 129; defensin, beta 129; hBD-29; prepro-beta-defensin 129 protein; UNQ5794/PRO19599
Gene Symbols DEFB129
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DEFB129. Recombinant protein control fragment (Product #RP-89631).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140881
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.