Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OHAUS™ Defender 6000 Extreme Washdown Scale Indicator
Shop All OHAUS Products
Click to view available options
Construction:
Polycarbonate, Stainless Steel (316)
Stainless Steel (316)
Model:
i-DT61PW AM
i-DT61PW AM Front Installation
i-DT61XWE AM
Description
- Applications: Weighing, Counting with APW optimization, Percent Weighing, Checkweighing, Checkcounting, Dynamic Weighing/Display Hold
- Display: 6-digit, 7-segment LCD display with white backlight, 45 mm high digits
- Operation: 6 D (LR20) Batteries (Included)
- Communication: Easy access IR communication kit (available as an accessory); GLP/GMP data output with real-time clock
- Construction: IP68/IP69K food grade polycarbonate housing and 316 stainless steel mounting bracket
- Design Features: 3-color check light bar, menu and key lock, selectable environmental and auto-print settings, stability indicator, overload/underload indicators, low battery indicator, auto shut-off, auto tare
Specifications
Specifications
| Battery Life | Up to 1,600 hours continuous use with backlight off |
| Construction | Polycarbonate, Stainless Steel (316) |
| Display Type | Backlit LCD |
| Model | i-DT61PW AM |
| Protection Class | IP68 immersion/IP69K washdown |
| Type | Bench Scales |
| Legal for Trade | Class III/IIIL/IIIHD |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction