Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Defensin 1/3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519228
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519228 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519228 Supplier Invitrogen™ Supplier No. PA519228
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.

Defensins are a family of microbicidal and cytotoxic peptides thought to be involved in host defense. They are abundant in the granules of neutrophils and also found in the epithelia of mucosal surfaces such as those of the intestine, respiratory tract, urinary tract, and vagina. Members of the defensin family are highly similar in protein sequence and distinguished by a conserved cysteine motif. The protein encoded by this gene, defensin, alpha 1, is found in the microbicidal granules of neutrophils and likely plays a role in phagocyte-mediated host defense. Several alpha defensin genes are clustered on chromosome 8. This gene differs from defensin, alpha 3 by only one amino acid. This gene and the gene encoding defensin, alpha 3 are both subject to copy number variation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Defensin 1/3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene DEFA1
Gene Accession No. P59665, P59666
Gene Alias alpha-defensin 1; Alpha-defensin 3; cryptdin-1; DEF1; DEF3; Defa1; DEFA1B; DEFA2; Defa27; Defa3; Defcr; Defcr1; Defcr3; Defcr-3; defensin 3, neutrophil-specific; defensin alpha 1; defensin alpha 3; defensin related cryptdin peptide 1; defensin, alpha 1; defensin, alpha 1, myeloid-related sequence; defensin, alpha 2; Defensin, alpha 3; defensin, alpha 3, neutrophil-specific; defensin, alpha, 27; defensin, alpha, 3; defensin-1; defensin-related cryptdin peptide; Defensin-related cryptdin-3; HNP-1; HNP-2; HNP3; HNP-3; HP 1-56; HP 3-56; HP1; HP-1; HP-2; HP3; HP-3; MRS; myeloid-related sequence; neutrophil defensin 1; Neutrophil defensin 2; Neutrophil defensin 3; neutrophil peptide 3
Gene Symbols DEFA1, DEFA3
Host Species Goat
Immunogen Synthetic peptide sequence (DESLAPKHPGSRKN) corresponding to the internal amino acids of DEFA1/3.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1667, 1668
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.