Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Defensin 5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5139755
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5139755 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5139755 Supplier Invitrogen™ Supplier No. PA5139755
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: ESLQERADEA TTQKQSGEDN QDLAISFAGN GLSALRTSGS QARATCYCRT GRCATRESLS GVCEISGRLY RLCCR Highest antigen sequence indentity to the following orthologs: Rat - 49%.

Defensins are a family of antimicrobial and cytotoxic peptides thought to be involved in host defense. They are abundant in the granules of neutrophils and also found in the epithelia of mucosal surfaces such as those of the intestine, respiratory tract, urinary tract, and vagina. Members of the defensin family are highly similar in protein sequence and distinguished by a conserved cysteine motif. Several of the alpha defensin genes appear to be clustered on chromosome 8. The protein encoded by this gene, defensin, alpha 5, is highly expressed in the secretory granules of Paneth cells of the ileum.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Defensin 5
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DEFA5
Gene Accession No. Q01523
Gene Alias DEF5; DEFA5; defensin 5; defensin alpha 5; Defensin, alpha 5; defensin, alpha 5, Paneth cell-specific; defensin, alpha 5, preproprotein; defensin-5; HD5; HD-5; HD5(20-94); HD5(23-94); HD5(29-94); HD5(56-94); HD5(63-94); MGC129728
Gene Symbols DEFA5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DEFA5. Recombinant protein control fragment (Product #RP-89636).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1670
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.