Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Defensin alpha-3 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5122038

 View more versions of this product

Catalog No. PIPA5122038


Only null left
Add to Cart
This item is not returnable. View return policy

Description

Description

Defensin alpha-3 Polyclonal Antibody for Western Blot, IHC (P)

Defensin alpha-1 is part of a family of antimicrobial and cytotoxic peptides thought to be involved in host defense. Defensins are abundant in the granules of neutrophils and also found in the epithelia of mucosal surfaces such as those of the intestine, respiratory tract, urinary tract, and vagina. Members of the defensin family are highly similar in protein sequence and distinguished by a conserved cysteine motif. The Defensin alpha-1 protein is encoded by the DFA1 gene and is found in the microbicidal granules of neutrophils and likely plays a role in phagocyte-mediated host defense. Several alpha defensin genes are clustered on chromosome 8, and this gene differs from defensin, alpha 3 by only one amino acid. The DFA1 gene and the gene encoding defensin, alpha 3 are both subject to copy number variation. Two transcript variants encoding different isoforms of Defensin alpha-1 have been found.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Defensin alpha-3
Polyclonal
Unconjugated
DEFA3
DEF3; DEFA3; defensin 3, neutrophil-specific; defensin alpha 3; Defensin, alpha 3; defensin, alpha 3, neutrophil-specific; HNP-2; HNP3; HNP-3; HP 3-56; HP-2; HP3; HP-3; Neutrophil defensin 2; Neutrophil defensin 3; neutrophil peptide 3
Rabbit
Antigen affinity chromatography, Protein A
RUO
1668
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
P59666
DEFA3
Recombinant protein Defensin Alpha 3, Neutrophil Specific. The antigen corresponds to amino acid range 21-94 of the target protein.
20 μL
Primary
Human, Pig
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.