Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Defensin alpha-3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567958
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567958 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567958 Supplier Invitrogen™ Supplier No. PA567958
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Defensin alpha-3 Polyclonal Antibody for Western Blot, IHC, IHC (P)

Defensin alpha-1 is part of a family of antimicrobial and cytotoxic peptides thought to be involved in host defense. Defensins are abundant in the granules of neutrophils and also found in the epithelia of mucosal surfaces such as those of the intestine, respiratory tract, urinary tract, and vagina. Members of the defensin family are highly similar in protein sequence and distinguished by a conserved cysteine motif. The Defensin alpha-1 protein is encoded by the DFA1 gene and is found in the microbicidal granules of neutrophils and likely plays a role in phagocyte-mediated host defense. Several alpha defensin genes are clustered on chromosome 8, and this gene differs from defensin, alpha 3 by only one amino acid. The DFA1 gene and the gene encoding defensin, alpha 3 are both subject to copy number variation. Two transcript variants encoding different isoforms of Defensin alpha-1 have been found.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Defensin alpha-3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 30% glycerol, 0.87% NaCl and 0.01% sodium azide; pH 7.3
Gene DEFA3
Gene Accession No. P59666
Gene Alias DEF3; DEFA3; defensin 3, neutrophil-specific; defensin alpha 3; Defensin, alpha 3; defensin, alpha 3, neutrophil-specific; HNP-2; HNP3; HNP-3; HP 3-56; HP-2; HP3; HP-3; Neutrophil defensin 2; Neutrophil defensin 3; neutrophil peptide 3
Gene Symbols DEFA3
Host Species Rabbit
Immunogen Synthesized peptide derived from human Defensin.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1668
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.