Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Defensin alpha 5 Antibody (8C8), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11060002R
Description
Defensin alpha 5 Monoclonal specifically detects Defensin alpha 5 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Defensin alpha 5 | |
Monoclonal | |
DyLight 550 | |
Q01523 | |
DEFA5 | |
Full-length human Defensin alpha 5 [Swiss-Prot# Q01523]. The propeptide is the full-length protein. In the mature form, it is cleaved at residue 63 (alanine). | |
Protein G purified | |
RUO | |
Primary | |
This is specific for HD5. It does not cross-react with HD6, lysozyme or sPLA2. | |
Store at 4C in the dark. | |
IgG2b |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
8C8 | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
DEF5defensin, alpha 5, preproprotein, defensin 5, Defensin, alpha 5, defensin, alpha 5, Paneth cell-specific, defensin-5, HD-5, MGC129728 | |
Mouse | |
10 kDa | |
0.1 mL | |
Immunology, Innate Immunity | |
1670 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction