Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Defensin beta 1 Antibody (1F5), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
| Antigen | Defensin beta 1 |
|---|---|
| Clone | 1F5 |
| Dilution | Flow Cytometry 1:50-1:200, ELISA |
| Applications | Flow Cytometry, ELISA |
| Classification | Monoclonal |
Description
Defensin beta 1 Monoclonal antibody specifically detects Defensin beta 1 in Human samples. It is validated for Flow Cytometry, ELISASpecifications
| Defensin beta 1 | |
| Flow Cytometry 1:50-1:200, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BD1, BD-1, beta-defensin 1, beta-defensin-1, DEFB-1, DEFB101, defensin, beta 1HBD-1, hBD-1, HBD1MGC51822 | |
| A synthesized peptide derived from Human Defensin beta 1 [UniProt P60022] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 1F5 | |
| Flow Cytometry, ELISA | |
| Unconjugated | |
| Rabbit | |
| Biologically Active Proteins | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 1672 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title