Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Defensin beta 1 Antibody (SR2223), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | Defensin beta 1 |
---|---|
Clone | SR2223 |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
Defensin beta 1 Monoclonal antibody specifically detects Defensin beta 1 in Human samples. It is validated for Western BlotSpecifications
Defensin beta 1 | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
BD1, BD-1, beta-defensin 1, beta-defensin-1, DEFB-1, DEFB101, defensin, beta 1HBD-1, hBD-1, HBD1MGC51822 | |
A synthesized peptide derived from human Defensin beta 1 (Uniprot #: P60022) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR2223 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Biologically Active Proteins | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
1672 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title