Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DEGS1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | DEGS1 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
DEGS1 Polyclonal specifically detects DEGS1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
DEGS1 | |
Polyclonal | |
Rabbit | |
Human | |
Cell migration-inducing gene 15 protein, Degenerative spermatocyte homolog 1, degenerative spermatocyte homolog 1, lipid desaturase (Drosophila), degenerative spermatocyte homolog, lipid desaturase (Drosophila), DEGS, Des-1, DES1degenerative spermatocyte homolog, lipid desaturase, EC 1.14, FADS7, membrane fatty acid (lipid) desaturase, Membrane lipid desaturase, MGC5079, migration-inducing gene 15 protein, MLDdihydroceramide desaturase, sphingolipid delta 4 desaturase, sphingolipid delta(4)-desaturase DES1 | |
DEGS1 | |
IgG | |
Affinity Purified |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
8560 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:YDNLPHYNSWIKVLYDFVMDDTISPYSRMKRHQKGEMVLE | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title