Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DEGS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA112912
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA112912 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA112912 Supplier Invitrogen™ Supplier No. PA112912
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control for this product is placenta lysate.

The product of the DEGS1 gene is a member of the membrane fatty acid (sphingolipid delta 4 ) desaturase family. Reported to function by inserting double bonds at delta 4 position in fatty acids. DEGS1 is an intracellular enzyme. Overexpression of this gene inhibited biosynthesis of the EGF receptor, suggesting a possible role of a fatty acid desaturase in regulation biosynthetic processing of the EGF receptor. Two splice variants have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DEGS1
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.08% sodium azide
Gene DEGS1
Gene Accession No. O09005, O15121, Q5XIF5
Gene Alias AA536663; Cell migration-inducing gene 15 protein; Degenerative spermatocyte homolog 1; degenerative spermatocyte homolog 1 (Drosophila); degenerative spermatocyte homolog 1, lipid desaturase; degenerative spermatocyte homolog, lipid desaturase; degenerative spermatocyte-like protein RDES; Degs; DEGS1; DEGS-1; delta 4-desaturase, sphingolipid 1; Delta(4)-desaturase; delta(4)-desaturase, sphingolipid 1; DES1; Des-1; dihydroceramide desaturase; dihydroceramide desaturase 1; dihydroceramide desaturase-1; DSH1; FADS7; fb81d06; hypothetical protein LOC507290; im:6909319; Mdes; membrane fatty acid (lipid) desaturase; membrane lipid desaturase; MGC5079; MIG15; migration-inducing gene 15 protein; MLD; sphingolipid 1; sphingolipid delta 4 desaturase; sphingolipid delta(4)-desaturase 1; sphingolipid delta(4)-desaturase DES1; wu:fa25h01; wu:fb81d06
Gene Symbols DEGS1
Host Species Rabbit
Immunogen Synthetic peptide derived from the human DEGS1 protein.
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13244, 58970, 8560
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.