Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fischer Technical Company™ Dehydrated Culture Media and Ingredients
SCI_ED

Catalog No. S012403 Shop All Fischer Technical Company Products
Change view
Click to view available options
Media Type:
Agar Powder
MacConkey Agar
Nutrient Agar
Nutrient Agar 1.5%
Nutrient Broth
Potato Dextrose Agar
Sabouraud Dextrose Agar
Tryptic Soy Agar
Quantity:
100 g
113 g
454 g
500 g
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Media Type Quantity
S012403 Potato Dextrose Agar 113 g
S71648A Nutrient Broth 454 g
S71614A Nutrient Agar 454 g
S71613A Nutrient Agar 113 g
S71666A Sabouraud Dextrose Agar 454 g
S70210A Agar Powder 100 g
S71646A Nutrient Agar 1.5% 454 g
S71609A MacConkey Agar 454 g
S71627A Tryptic Soy Agar 454 g
S71659A Potato Dextrose Agar 454 g
S70213A Agar Powder 500 g
S012404 Nutrient Broth 113 g
S012406 Tryptic Soy Agar 113 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. S012403 Supplier Fischer Technical Company™ Supplier No. CD149A

Essential agents for preparing culture media.

Successful bacteriological work begins with high-quality media.

  • Agar, a solidifying agent, is an essential component of many media
  • Available in a variety of formulations and bottle sizes
  • For use in preparing plated media, tubed media, bottled media, and slants

Specifications

Detects Bacteria and Fungi
Form Dehydrated Powder
Format Bottled
Manufacturing Origin US with Globally Sourced Materials
Media Type Potato Dextrose Agar
Product Type Microbiology Media
Sterility Non-sterile
Target Organism Class Bacteria, Fungi, Yeast, Molds
Test Type Microbiological Tests
For Use With (Application) Microbiological Testing
Quantity 113 g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.